ENA405194159 Enamine

2-[2-(phenylformamido)ethoxy]acetic acid

  • MDL number MFCD21933516
  • Cas Number : 142489-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133144

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133144
Availability details for
Quantity
  • Availability
  • Add to Cart