ENA405194161 Enamine

1-(benzyloxy)-4-(2-nitrobut-1-en-1-yl)benzene

  • MDL number MFCD25970319
  • Cas Number : 71488-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133154

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133154
Availability details for
Quantity
  • Availability
  • Add to Cart