ENA405194164 Enamine

4-bromo-3-(chlorosulfonyl)-5-fluorobenzoic acid

  • MDL number MFCD16084850
  • Cas Number : 1242338-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213...215
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107777

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107777
Availability details for
Quantity
  • Availability
  • Add to Cart