ENA405194171 Enamine

1-fluoro-2-nitro-3-(propan-2-yloxy)benzene

  • MDL number MFCD13689267
  • Cas Number : 1233951-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133177

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133177
Availability details for
Quantity
  • Availability
  • Add to Cart