ENA405194172 Enamine

2,3-dimethyloxolane-3-carboxylic acid

  • MDL number MFCD25970323
  • Cas Number : 1461715-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133198
Availability details for
Quantity
  • Availability
  • Add to Cart