ENA405194173 Enamine

tert-butyl 3-(2-hydroxyethyl)azetidine-1-carboxylate

  • MDL number MFCD10699280
  • Cas Number : 152537-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133199
Availability details for
Quantity
  • Availability
  • Add to Cart