ENA405194177 Enamine

2,5-dichloro-4-methylpyridine-3-carbonitrile

  • MDL number MFCD07368156
  • Cas Number : 101320-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133208
Availability details for
Quantity
  • Availability
  • Add to Cart