ENA405194178 Enamine

4-(4-methoxyphenyl)-2-methyl-4-oxobutanenitrile

  • MDL number MFCD11893475
  • Cas Number : 1181379-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-109798

Safety Information

msds https://www.enaminestore.com/catalog/EN300-109798
Availability details for
Quantity
  • Availability
  • Add to Cart