ENA405194187 Enamine

2-chloro-4-[(4-chloro-1H-pyrazol-1-yl)methyl]pyridine

  • MDL number MFCD16765174
  • Cas Number : 1248056-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133231
Availability details for
Quantity
  • Availability
  • Add to Cart