ENA405194190 Enamine

2,6-diaminohex-4-ynoic acid dihydrochloride

  • MDL number MFCD25970331
  • Cas Number : 65579-76-2
  • Molecular Weight : 215.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 192-194
Iupac Name 2,6-diamino-4-hexynoic acid dihydrochloride
Inchi Code 1S/C6H10N2O2.2ClH/c7-4-2-1-3-5(8)6(9)10;;/h5H,3-4,7-8H2,(H,9,10);2*1H
Inchi Key IHEPKHXGAAYYQN-UHFFFAOYSA-N

Description

Melting Point 192-194
Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-133236

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-133236
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart