ENA405194194 Enamine

2-{5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyridin-3-yl}ethan-1-amine dihydrochloride

  • MDL number MFCD25970335
  • Cas Number : 1461715-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133240

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133240
Availability details for
Quantity
  • Availability
  • Add to Cart