ENA405194196 Enamine

bicyclo[2.2.1]heptane-1-carboxylic acid

  • MDL number MFCD03413212
  • Cas Number : 18720-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110...112
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113199

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-113199
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart