ENA405194202 Enamine

3,3,5-trimethylpiperidine

  • MDL number MFCD19218107
  • Cas Number : 37832-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133280
Availability details for
Quantity
  • Availability
  • Add to Cart