ENA405194205 Enamine

3-methylbut-3-en-2-ol

  • MDL number MFCD00060892
  • Cas Number : 10473-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114325

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114325
Availability details for
Quantity
  • Availability
  • Add to Cart