ENA405194208 Enamine

1-cyclohexyl-1H-1,2,4-triazol-3-amine

  • MDL number MFCD12804863
  • Cas Number : 1179738-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-133288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133288
Availability details for
Quantity
  • Availability
  • Add to Cart