ENA405194211 Enamine

2,6-dimethyloxan-4-ol

  • MDL number MFCD18089272
  • Cas Number : 41866-70-0
  • Molecular Weight : 130.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-49
Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-114337

Safety Information

signalword Danger
pcodes P280,P305+P351+P338,P310
MSDS https://www.enaminestore.com/catalog/EN300-114337
pictograms GHS05
hstatements H318
Availability details for
Quantity
  • Availability
  • Add to Cart