ENA405194214 Enamine

(2-methoxypyrimidin-4-yl)methanol

  • MDL number MFCD18642499
  • Cas Number : 38696-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133304
Availability details for
Quantity
  • Availability
  • Add to Cart