ENA405194215 Enamine

3-methyl-5-nitroisoquinoline

  • MDL number MFCD11110422
  • Cas Number : 18222-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-115241

Safety Information

msds https://www.enaminestore.com/catalog/EN300-115241
Availability details for
Quantity
  • Availability
  • Add to Cart