ENA405194231 Enamine

ethyl 5-amino-2-(phenylsulfanyl)benzoate

  • MDL number MFCD13343766
  • Cas Number : 1308601-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133384

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133384
Availability details for
Quantity
  • Availability
  • Add to Cart