ENA405194234 Enamine

2-(piperidine-1-sulfonyl)acetonitrile

  • MDL number MFCD09041547
  • Cas Number : 926253-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133389

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133389
Availability details for
Quantity
  • Availability
  • Add to Cart