ENA405194236 Enamine

4-(chloromethyl)-2,6-dimethylpyrimidine

  • MDL number MFCD16868258
  • Cas Number : 89966-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133391
Availability details for
Quantity
  • Availability
  • Add to Cart