ENA405194239 Enamine

7-fluoro-6-methyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD16658724
  • Cas Number : 1037206-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133395
Availability details for
Quantity
  • Availability
  • Add to Cart