ENA405194240 Enamine

5-bromonaphthalene-1-carbonitrile

  • MDL number MFCD17012496
  • Cas Number : 129278-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-117502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-117502
Availability details for
Quantity
  • Availability
  • Add to Cart