ENA405194241 Enamine

5-chloro-6-(cyclopropylmethoxy)pyridine-3-carboxylic acid

  • MDL number MFCD11136671
  • Cas Number : 1019493-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133397

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133397
Availability details for
Quantity
  • Availability
  • Add to Cart