ENA405194244 Enamine

4-[4-(trifluoromethyl)phenyl]butanoic acid

  • MDL number MFCD06810491
  • Cas Number : 136295-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-118524

Safety Information

msds https://www.enaminestore.com/catalog/EN300-118524
Availability details for
Quantity
  • Availability
  • Add to Cart