ENA405194245 Enamine

2-methyl-N'-(5-nitropyridin-2-yl)propanehydrazide

  • MDL number MFCD25970349
  • Cas Number : 1329296-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212...214
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133410

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133410
Availability details for
Quantity
  • Availability
  • Add to Cart