ENA405194247 Enamine

4-cyclohexylmorpholine-2-carbonitrile

  • MDL number MFCD25970351
  • Cas Number : 1461714-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133436

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133436
Availability details for
Quantity
  • Availability
  • Add to Cart