ENA405194248 Enamine

tert-butyl 3-(aminomethyl)-3-methylpiperidine-1-carboxylate

  • MDL number MFCD12408579
  • Cas Number : 1158759-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133441

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133441
Availability details for
Quantity
  • Availability
  • Add to Cart