ENA405194252 Enamine

tert-butyl 4-phenyl-4-[(2,2,2-trifluoroethyl)carbamoyl]piperidine-1-carboxylate

  • MDL number MFCD25970353
  • Cas Number : 1461709-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133446

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133446
Availability details for
Quantity
  • Availability
  • Add to Cart