ENA405194253 Enamine

2-chloromethanesulfonyl-2-methylpropane

  • MDL number MFCD00092208
  • Cas Number : 24824-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-85
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133449
Availability details for
Quantity
  • Availability
  • Add to Cart