ENA405194257 Enamine

3-(methoxymethyl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD14690991
  • Cas Number : 115443-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138...140
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133459

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133459
Availability details for
Quantity
  • Availability
  • Add to Cart