ENA405194270 Enamine

1-propylazetidin-3-ol

  • MDL number MFCD18308893
  • Cas Number : 1340301-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133487
Availability details for
Quantity
  • Availability
  • Add to Cart