ENA405194272 Enamine

2-(2-oxo-2,3-dihydro-1H-indol-6-yl)acetic acid

  • MDL number MFCD01110749
  • Cas Number : 101566-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-253
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133489
Availability details for
Quantity
  • Availability
  • Add to Cart