ENA405194276 Enamine

1-amino-3-methyl-1,3-diazinan-2-one

  • MDL number MFCD25970359
  • Cas Number : 1461709-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133493
Availability details for
Quantity
  • Availability
  • Add to Cart