ENA405194280 Enamine

4-(4-aminobenzamido)-N-methylbenzamide

  • MDL number MFCD12594692
  • Cas Number : 1156046-70-6
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-133498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133498
Availability details for
Quantity
  • Availability
  • Add to Cart