ENA405194282 Enamine

2-(oxolan-3-yloxy)pyridine-4-carboxylic acid

  • MDL number MFCD18259827
  • Cas Number : 1211533-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122851

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122851
Availability details for
Quantity
  • Availability
  • Add to Cart