ENA405194288 Enamine

2-(3-iodopropoxy)oxane

  • MDL number MFCD25964111
  • Cas Number : 52103-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133541

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133541
Availability details for
Quantity
  • Availability
  • Add to Cart