ENA405194294 Enamine

1-benzoylpiperidin-3-ol

  • MDL number MFCD03790922
  • Cas Number : 67452-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133626

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133626
Availability details for
Quantity
  • Availability
  • Add to Cart