ENA405194304 Enamine

2-(2-methyloxan-4-yl)ethan-1-amine

  • MDL number MFCD19593680
  • Cas Number : 1343285-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133656

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133656
Availability details for
Quantity
  • Availability
  • Add to Cart