ENA405194307 Enamine

3,5-dimethyl-1-(2-methylpropyl)-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD12192675
  • Cas Number : 1156602-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-133659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133659
Availability details for
Quantity
  • Availability
  • Add to Cart