ENA405194315 Enamine

1-(2,5-difluoro-4-methoxyphenyl)-5-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD25970367
  • Cas Number : 1461709-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106...108
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133674

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133674
Availability details for
Quantity
  • Availability
  • Add to Cart