ENA405194320 Enamine

(3-methoxyphenyl)methanesulfonamide

  • MDL number MFCD16671946
  • Cas Number : 89782-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133680

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133680
Availability details for
Quantity
  • Availability
  • Add to Cart