ENA405194336 Enamine

methyl 2-(2-fluoro-4-nitrophenyl)propanoate

  • MDL number MFCD23096898
  • Cas Number : 104784-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133708
Availability details for
Quantity
  • Availability
  • Add to Cart