ENA405194352 Enamine

N-(pyrrolidin-3-yl)propanamide

  • MDL number MFCD12085764
  • Cas Number : 1154969-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133755

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133755
Availability details for
Quantity
  • Availability
  • Add to Cart