ENA405194356 Enamine

2-(3,4-dimethoxyphenyl)-2-ethylbutanenitrile

  • MDL number MFCD12846537
  • Cas Number : 92500-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133763
Availability details for
Quantity
  • Availability
  • Add to Cart