ENA405194360 Enamine

4-(dimethylamino)pyridine-3-carboxylic acid

  • MDL number MFCD20654938
  • Cas Number : 83039-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133767

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133767
Availability details for
Quantity
  • Availability
  • Add to Cart