ENA405194373 Enamine

ethyl 1-benzoylpyrrolidine-2-carboxylate

  • MDL number MFCD15485360

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133811

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133811
Availability details for
Quantity
  • Availability
  • Add to Cart