ENA405194379 Enamine

6-chloro-3-(propan-2-yl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03001443
  • Cas Number : 887581-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190...192
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133825

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133825
Availability details for
Quantity
  • Availability
  • Add to Cart