ENA405194380 Enamine

3,5-dimethyl-1-(2-methylpropyl)-1H-pyrazole-4-sulfonamide

  • MDL number MFCD14657994
  • Cas Number : 1251037-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133826

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133826
Availability details for
Quantity
  • Availability
  • Add to Cart