ENA405194389 Enamine

5-(piperidin-4-yl)-2,3-dihydro-1H-pyrazol-3-one dihydrochloride

  • MDL number MFCD25970262
  • Cas Number : 1461706-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-127726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-127726
Availability details for
Quantity
  • Availability
  • Add to Cart